5-(2-Methoxyphenyl)thiophene-2-carbaldehyde

Lieferant: Apollo Scientific

OR470275-5G
APOSOR470275-5GEA 1410 EUR
APOSOR470275-5G
5-(2-Methoxyphenyl)thiophene-2-carbaldehyde
5-(2-Methoxyphenyl)thiophene-2-carbaldehyde
Formel: C₁₂H₁₀O₂S
Molecular Weight: 218,272 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410189
CAS-Nummer: 479243-27-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR