[6-(4-Methoxyphenyl)pyridin-3-yl]methanol

Lieferant: Apollo Scientific

OR470280-5G
APOSOR470280-5GEA 1540 EUR
APOSOR470280-5G
[6-(4-Methoxyphenyl)pyridin-3-yl]methanol
[6-(4-Methoxyphenyl)pyridin-3-yl]methanol
Formel: C₁₃H₁₃NO₂
Molecular Weight: 215,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410445
CAS-Nummer: 887974-96-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR