5-(2-Chlorophenyl)-3-pyridinecarbaldehyde

Lieferant: Apollo Scientific

OR470281-5G
APOSOR470281-5GEA 1730 EUR
APOSOR470281-5G
5-(2-Chlorophenyl)-3-pyridinecarbaldehyde
5-(2-Chlorophenyl)-3-pyridinecarbaldehyde
Formel: C₁₂H₈ClNO
Molecular Weight: 217,651 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410222
CAS-Nummer: 855301-00-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR