Ethyl 3-(1,4-Benzodioxan-2-yl)-3-oxopropanoate

Lieferant: Apollo Scientific

OR470283-5G
APOSOR470283-5GEA 1120 EUR
APOSOR470283-5G
Ethyl 3-(1,4-Benzodioxan-2-yl)-3-oxopropanoate
Ethyl 3-(1,4-Benzodioxan-2-yl)-3-oxopropanoate
Formel: C₁₃H₁₄O₅
Molecular Weight: 250,247 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07370064
CAS-Nummer: 889955-17-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR