[5-(4-Methoxyphenyl)-3-pyridyl]methanol

Lieferant: Apollo Scientific

OR470284-5G
APOSOR470284-5GEA 1540 EUR
APOSOR470284-5G
[5-(4-Methoxyphenyl)-3-pyridyl]methanol
[5-(4-Methoxyphenyl)-3-pyridyl]methanol
Formel: C₁₃H₁₃NO₂
Molecular Weight: 215,248 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410316
CAS-Nummer: 887974-21-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR