3-[3-[2-(2-Tetrahydropyranyloxy)ethyl]ureido]thiophene-2-carboxylic acid
Lieferant: Apollo Scientific
OR470286-5G
APOSOR470286-5GEA
1540
EUR
APOSOR470286-5G
3-[3-[2-(2-Tetrahydropyranyloxy)ethyl]ureido]thiophene-2-carboxylic acid
3-[3-[2-(2-Tetrahydropyranyloxy)ethyl]ureido]thiophene-2-carboxylic acid
Formel:
C₁₃H₁₈N₂O₅S Molecular Weight: 314.357 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18433028 CAS-Nummer: 1265892-01-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...