Methyl 3-(1-Benzyl-1,2,5,6-tetrahydro-3-pyridyl)propanoate

Lieferant: Apollo Scientific

OR470287-1G
APOSOR470287-1GEA 1210 EUR
APOSOR470287-1G
Methyl 3-(1-Benzyl-1,2,5,6-tetrahydro-3-pyridyl)propanoate
Methyl 3-(1-Benzyl-1,2,5,6-tetrahydro-3-pyridyl)propanoate
Formel: C₁₆H₂₁NO₂
Molecular Weight: 259,343 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22423105
CAS-Nummer: 1398534-58-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR