Methyl 3-(1-Benzyl-1,2,3,6-tetrahydro-4-pyridyl)propanoate

Lieferant: Apollo Scientific

OR470288-1G
APOSOR470288-1GEA 1210 EUR
APOSOR470288-1G
Methyl 3-(1-Benzyl-1,2,3,6-tetrahydro-4-pyridyl)propanoate
Methyl 3-(1-Benzyl-1,2,3,6-tetrahydro-4-pyridyl)propanoate
Formel: C₁₆H₂₁NO₂
Molecular Weight: 259,343 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22423106
CAS-Nummer: 1398534-62-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR