4-(Aminomethyl)-1-benzyl-1,2,3,6-tetrahydropyridine

Lieferant: Apollo Scientific

Synonyme: (1-Benzyl-1,2,3,6-tetrahydropyridin-4-yl)methanamine

OR470289-1G
APOSOR470289-1GEA 1210 EUR
APOSOR470289-1G
4-(Aminomethyl)-1-benzyl-1,2,3,6-tetrahydropyridine
4-(Aminomethyl)-1-benzyl-1,2,3,6-tetrahydropyridine
Formel: C₁₃H₁₈N₂
Molecular Weight: 202,295 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22423107
CAS-Nummer: 153196-51-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR