3-(Aminomethyl)-1-benzyl-1,2,5,6-tetrahydropyridine

Lieferant: Apollo Scientific

OR470290-1G
APOSOR470290-1GEA 1210 EUR
APOSOR470290-1G
3-(Aminomethyl)-1-benzyl-1,2,5,6-tetrahydropyridine
3-(Aminomethyl)-1-benzyl-1,2,5,6-tetrahydropyridine
Formel: C₁₃H₁₈N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22423108
CAS-Nummer: 124257-63-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR