Methyl 3-Chloro-4-piperazinobenzoate
Lieferant: Apollo Scientific
OR470292-5G
OR470292-1G
APOSOR470292-5GEA
804
EUR
APOSOR470292-5G
APOSOR470292-1G
Methyl 3-Chloro-4-piperazinobenzoate
Methyl 3-Chloro-4-piperazinobenzoate
Formel:
C₁₂H₁₅ClN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD19442776 CAS-Nummer: 234082-16-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...