Methyl 3-Chloro-4-piperazinobenzoate

Lieferant: Apollo Scientific

OR470292-5G OR470292-1G
APOSOR470292-5GEA 804 EUR
APOSOR470292-5G APOSOR470292-1G
Methyl 3-Chloro-4-piperazinobenzoate
Methyl 3-Chloro-4-piperazinobenzoate
Formel: C₁₂H₁₅ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD19442776
CAS-Nummer: 234082-16-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR