N-Cbz-2-morpholinecarboxylic acid
Lieferant: Apollo Scientific
OR470293-5G
APOSOR470293-5GEA
0
EUR
APOSOR470293-5G
N-Cbz-2-morpholinecarboxylic acid
N-Cbz-2-morpholinecarboxylic acid
Formel:
C₁₃H₁₅NO₅ Lagertemperatur: Raumtemperatur |
MDL:
MFCD16223609 CAS-Nummer: 135782-21-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...