Methyl 3-chloro-4-morpholinobenzoate
Lieferant: Apollo Scientific
OR470294-5G
APOSOR470294-5GEA
713
EUR
APOSOR470294-5G
Methyl 3-chloro-4-morpholinobenzoate
Methyl 3-chloro-4-morpholinobenzoate
Formel:
C₁₂H₁₄ClNO₃ Molecular Weight: 255,697 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19442778 CAS-Nummer: 1314406-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...