3-Acetamido-4-methylpyridazine

Lieferant: Apollo Scientific

OR470296-5G
APOSOR470296-5GEA 1540 EUR
APOSOR470296-5G
3-Acetamido-4-methylpyridazine
3-Acetamido-4-methylpyridazine
Formel: C₇H₉N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD19442780
CAS-Nummer: 1314406-37-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR