3-Acetamido-5-methylpyridazine

Lieferant: Apollo Scientific

OR470297-5G
APOSOR470297-5GEA 1540 EUR
APOSOR470297-5G
3-Acetamido-5-methylpyridazine
3-Acetamido-5-methylpyridazine
Formel: C₇H₉N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD19442781
CAS-Nummer: 1314406-52-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR