2-Methyl-1,3-dioxolane-2-acetamide

Lieferant: Apollo Scientific

OR470298-5G
APOSOR470298-5GEA 916 EUR
APOSOR470298-5G
2-Methyl-1,3-dioxolane-2-acetamide
2-Methyl-1,3-dioxolane-2-acetamide
Formel: C₆H₁₁NO₃
Molecular Weight: 145,156 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19442782
CAS-Nummer: 70829-14-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR