Methyl 2,6-Dihydroxypyridine-4-carboxylate hydrochloride

Lieferant: Apollo Scientific

OR470299-5G
APOSOR470299-5GEA 397 EUR
APOSOR470299-5G
Methyl 2,6-Dihydroxypyridine-4-carboxylate hydrochloride
Methyl 2,6-Dihydroxypyridine-4-carboxylate hydrochloride
Formel: C₇H₇NO₄·ClH
Molecular Weight: 205,596 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19442784
CAS-Nummer: 1314406-42-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR