2,6-Dihydroxypyridine-4-carboxamide

Lieferant: Apollo Scientific

OR470300-5G
APOSOR470300-5GEA 448 EUR
APOSOR470300-5G
2,6-Dihydroxypyridine-4-carboxamide
2,6-Dihydroxypyridine-4-carboxamide
Formel: C₆H₆N₂O₃
Molecular Weight: 154,123 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19442785
CAS-Nummer: 14533-64-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR