Diethyl 2-(6-Methoxy-3-pyridyl)malonate

Lieferant: Apollo Scientific

OR470301-5G
APOSOR470301-5GEA 997 EUR
APOSOR470301-5G
Diethyl 2-(6-Methoxy-3-pyridyl)malonate
Diethyl 2-(6-Methoxy-3-pyridyl)malonate
Formel: C₁₃H₁₇NO₅
Lagertemperatur: Raumtemperatur
MDL: MFCD19442788
CAS-Nummer: 902130-84-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR