2-[(Cyclopropylcarbonyl)amino]benzoic acid
Lieferant: Apollo Scientific
OR470307-5G
APOSOR470307-5GEA
734
EUR
APOSOR470307-5G
2-[(Cyclopropylcarbonyl)amino]benzoic acid
2-[(Cyclopropylcarbonyl)amino]benzoic acid
Formel:
C₁₁H₁₁NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD02602218 CAS-Nummer: 40057-21-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...