4-Chloro-2-(methylsulphonamido)benzoic acid
Lieferant: Apollo Scientific
OR470321-5G
APOSOR470321-5GEA
734
EUR
APOSOR470321-5G
4-Chloro-2-(methylsulphonamido)benzoic acid
4-Chloro-2-(methylsulphonamido)benzoic acid
Formel:
C₈H₈ClNO₄S Molecular Weight: 249,671 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03975839 CAS-Nummer: 158579-89-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...