3-(Fmoc-amino)-3-(3-tert-butoxyphenyl)propanoic acid

Lieferant: Apollo Scientific

OR470325-5G
APOSOR470325-5GEA 1540 EUR
APOSOR470325-5G
3-(Fmoc-amino)-3-(3-tert-butoxyphenyl)propanoic acid
3-(Fmoc-amino)-3-(3-tert-butoxyphenyl)propanoic acid
Formel: C₂₈H₂₉NO₅
Molecular Weight: 459,534 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02683182
CAS-Nummer: 372144-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR