2-Amino-2-(3,5-dichlorophenyl)acetic acid hydrochloride

Lieferant: Apollo Scientific

OR470326-5G
APOSOR470326-5GEA 1330 EUR
APOSOR470326-5G
2-Amino-2-(3,5-dichlorophenyl)acetic acid hydrochloride
2-Amino-2-(3,5-dichlorophenyl)acetic acid hydrochloride
Formel: C₈H₇Cl₂NO₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD06202321
CAS-Nummer: 1137014-87-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR