6-(4-Methoxyphenyl)nicotinic acid

Lieferant: Apollo Scientific

OR470328-5G
APOSOR470328-5GEA 1540 EUR
APOSOR470328-5G
6-(4-Methoxyphenyl)nicotinic acid
6-(4-Methoxyphenyl)nicotinic acid
Formel: C₁₃H₁₁NO₃
Molecular Weight: 229.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410402
CAS-Nummer: 223127-23-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR