5-(4-Bromophenyl)nicotinic acid

Lieferant: Apollo Scientific

OR470330-5G
APOSOR470330-5GEA 1410 EUR
APOSOR470330-5G
5-(4-Bromophenyl)nicotinic acid
5-(4-Bromophenyl)nicotinic acid
Formel: C₁₂H₈BrNO₂
Molecular Weight: 278.101 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410263
CAS-Nummer: 887973-36-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR