1-(Boc-amino)-3-bromo-1-(3-chlorophenyl)propane
Lieferant: Apollo Scientific
OR470331-5G
APOSOR470331-5GEA
1540
EUR
APOSOR470331-5G
1-(Boc-amino)-3-bromo-1-(3-chlorophenyl)propane
1-(Boc-amino)-3-bromo-1-(3-chlorophenyl)propane
Formel:
C₁₄H₁₉BrClNO₂ Molecular Weight: 348,663 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06410609 CAS-Nummer: 924817-77-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...