2-Amino-6-chloro-3-methylbenzoic acid

Lieferant: Apollo Scientific

OR470333-5G
APOSOR470333-5GEA 1120 EUR
APOSOR470333-5G
2-Amino-6-chloro-3-methylbenzoic acid
2-Amino-6-chloro-3-methylbenzoic acid
Formel: C₈H₈ClNO₂
Molecular Weight: 185,608 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06409109
CAS-Nummer: 857005-81-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR