6-(2-Methoxyphenyl)nicotinic acid

Lieferant: Apollo Scientific

OR470336-1G OR470336-5G OR470336-250MG
APOSOR470336-1GEA 493 EUR
APOSOR470336-1G APOSOR470336-5G APOSOR470336-250MG
6-(2-Methoxyphenyl)nicotinic acid
6-(2-Methoxyphenyl)nicotinic acid
Formel: C₁₃H₁₁NO₃
Molecular Weight: 229.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410400
CAS-Nummer: 887976-03-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR