4-(Benzylamino)tetrahydrothiopyran
Lieferant: Apollo Scientific
OR470341-5G
APOSOR470341-5GEA
997
EUR
APOSOR470341-5G
4-(Benzylamino)tetrahydrothiopyran
4-(Benzylamino)tetrahydrothiopyran
Formel:
C₁₂H₁₇NS Molecular Weight: 207,335 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06410637 CAS-Nummer: 887978-08-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...