5-(3-Methoxyphenyl)-2-thiophenecarbaldehyde
Lieferant: Apollo Scientific
OR470343-5G
APOSOR470343-5GEA
1540
EUR
APOSOR470343-5G
5-(3-Methoxyphenyl)-2-thiophenecarbaldehyde
5-(3-Methoxyphenyl)-2-thiophenecarbaldehyde
Formel:
C₁₂H₁₀O₂S Molecular Weight: 218,272 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06410190 CAS-Nummer: 249504-37-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...