[5-(2-Thienyl)-3-pyridyl]methanol

Lieferant: Apollo Scientific

Synonyme: (5-(Thiophen-2-yl)pyridin-3-yl)methanol

OR470344-1G
APOSOR470344-1GEA 713 EUR
APOSOR470344-1G
[5-(2-Thienyl)-3-pyridyl]methanol
[5-(2-Thienyl)-3-pyridyl]methanol
Formel: C₁₀H₉NOS
Molecular Weight: 191,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410298
CAS-Nummer: 393861-00-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR