2-Amino-2-[6-(4-chlorophenyl)-3-pyridyl]acetic acid hydrochloride

Lieferant: Apollo Scientific

OR470345-5G
APOSOR470345-5GEA 1840 EUR
APOSOR470345-5G
2-Amino-2-[6-(4-chlorophenyl)-3-pyridyl]acetic acid hydrochloride
2-Amino-2-[6-(4-chlorophenyl)-3-pyridyl]acetic acid hydrochloride
Formel: C₁₃H₁₁ClN₂O₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD07371467
CAS-Nummer: 1137666-85-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR