4-(Aminomethyl)-4-p-tolylcyclohexanol

Lieferant: Apollo Scientific

OR470346-5G
APOSOR470346-5GEA 1840 EUR
APOSOR470346-5G
4-(Aminomethyl)-4-p-tolylcyclohexanol
4-(Aminomethyl)-4-p-tolylcyclohexanol
Formel: C₁₄H₂₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD06410565
CAS-Nummer: 887978-24-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR