Fmoc-2-amino-5-bromobenzoic acid

Lieferant: Apollo Scientific

OR470347-5G
APOSOR470347-5GEA 601 EUR
APOSOR470347-5G
Fmoc-2-amino-5-bromobenzoic acid
Fmoc-2-amino-5-bromobenzoic acid
Formel: C₂₂H₁₆BrNO₄
Molecular Weight: 438.29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02682151
CAS-Nummer: 183871-04-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR