N-(4-Tetrahydropyranylmethyl)benzylamine

Lieferant: Apollo Scientific

OR470348-5G
APOSOR470348-5GEA 1120 EUR
APOSOR470348-5G
N-(4-Tetrahydropyranylmethyl)benzylamine
N-(4-Tetrahydropyranylmethyl)benzylamine
Formel: C₁₃H₁₉NO
Molecular Weight: 205,296 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07371441
CAS-Nummer: 887978-20-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR