3-(Boc-amino)-3-(4-tetrahydrothiopyranyl)-1-propanol

Lieferant: Apollo Scientific

OR470349-1G
APOSOR470349-1GEA 713 EUR
APOSOR470349-1G
3-(Boc-amino)-3-(4-tetrahydrothiopyranyl)-1-propanol
3-(Boc-amino)-3-(4-tetrahydrothiopyranyl)-1-propanol
Formel: C₁₃H₂₅NO₃S
Molecular Weight: 275,408 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115545
CAS-Nummer: 898405-01-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR