Fmoc-2-amino-5-methoxybenzoic acid

Lieferant: Apollo Scientific

OR470350-5G
APOSOR470350-5GEA 1120 EUR
APOSOR470350-5G
Fmoc-2-amino-5-methoxybenzoic acid
Fmoc-2-amino-5-methoxybenzoic acid
Formel: C₂₃H₁₉NO₅
Molecular Weight: 389,41 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02682215
CAS-Nummer: 332121-93-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR