5-Methyl-2-(phenylethynyl)pyridine

Lieferant: Apollo Scientific

OR470351-1G
APOSOR470351-1GEA 137 EUR
APOSOR470351-1G
5-Methyl-2-(phenylethynyl)pyridine
5-Methyl-2-(phenylethynyl)pyridine
Formel: C₁₄H₁₁N
Molecular Weight: 193,244 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18205120
CAS-Nummer: 124300-38-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR