2-Hydroxy-5-pentylbenzoic acid
Lieferant: Apollo Scientific
OR470353-5G
APOSOR470353-5GEA
1120
EUR
APOSOR470353-5G
2-Hydroxy-5-pentylbenzoic acid
2-Hydroxy-5-pentylbenzoic acid
Formel:
C₁₂H₁₆O₃ Molecular Weight: 208,254 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11203636 CAS-Nummer: 28488-46-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...