3-(Boc-amino)-3-(3-chlorophenyl)-1-propanol
Lieferant: Apollo Scientific
OR470354-5G
APOSOR470354-5GEA
1330
EUR
APOSOR470354-5G
3-(Boc-amino)-3-(3-chlorophenyl)-1-propanol
3-(Boc-amino)-3-(3-chlorophenyl)-1-propanol
Formel:
C₁₄H₂₀ClNO₃ Molecular Weight: 285,766 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18907505 CAS-Nummer: 1314406-54-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...