Ethyl 3-Oxo-3-(6-quinolyl)propanoate
Lieferant: Apollo Scientific
OR470355-5G
APOSOR470355-5GEA
1120
EUR
APOSOR470355-5G
Ethyl 3-Oxo-3-(6-quinolyl)propanoate
Ethyl 3-Oxo-3-(6-quinolyl)propanoate
Formel:
C₁₄H₁₃NO₃ Molecular Weight: 243,258 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07370067 CAS-Nummer: 858646-09-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...