5-(2-Methylphenyl)-2-thiophenecarbaldehyde

Lieferant: Apollo Scientific

OR470356-5G
APOSOR470356-5GEA 1540 EUR
APOSOR470356-5G
5-(2-Methylphenyl)-2-thiophenecarbaldehyde
5-(2-Methylphenyl)-2-thiophenecarbaldehyde
Formel: C₁₂H₁₀OS
Lagertemperatur: Raumtemperatur
MDL: MFCD06410187
CAS-Nummer: 886509-95-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR