Ethyl 3-Oxo-4-(1-Boc-4-piperidyl)butyrate

Lieferant: Apollo Scientific

OR470357-5G
APOSOR470357-5GEA 1410 EUR
APOSOR470357-5G
Ethyl 3-Oxo-4-(1-Boc-4-piperidyl)butyrate
Ethyl 3-Oxo-4-(1-Boc-4-piperidyl)butyrate
Formel: C₁₆H₂₇NO₅
Molecular Weight: 313,389 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07370059
CAS-Nummer: 916791-39-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR