3-(Boc-amino)-6,6-dichlorobicyclo[3.1.0]hexane-3-carboxylic acid

Lieferant: Apollo Scientific

OR470360-1G
APOSOR470360-1GEA 997 EUR
APOSOR470360-1G
3-(Boc-amino)-6,6-dichlorobicyclo[3.1.0]hexane-3-carboxylic acid
3-(Boc-amino)-6,6-dichlorobicyclo[3.1.0]hexane-3-carboxylic acid
Formel: C₁₂H₁₇Cl₂NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD04115621
CAS-Nummer: 1134759-39-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR