5-(2-Chlorophenyl)nicotinic acid

Lieferant: Apollo Scientific

OR470362-5G
APOSOR470362-5GEA 1120 EUR
APOSOR470362-5G
5-(2-Chlorophenyl)nicotinic acid
5-(2-Chlorophenyl)nicotinic acid
Formel: C₁₂H₈ClNO₂
Molecular Weight: 233.65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410264
CAS-Nummer: 893735-02-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR