6-(3,5-Dichlorophenyl)-3-pyridinecarbaldehyde

Lieferant: Apollo Scientific

OR470364-1G
APOSOR470364-1GEA 1120 EUR
APOSOR470364-1G
6-(3,5-Dichlorophenyl)-3-pyridinecarbaldehyde
6-(3,5-Dichlorophenyl)-3-pyridinecarbaldehyde
Formel: C₁₂H₇Cl₂NO
Molecular Weight: 252,096 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115466
CAS-Nummer: 898796-01-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR