4-(Aminomethyl)-4-(3,4-dichlorophenyl)cyclohexanol

Lieferant: Apollo Scientific

OR470367-5G
APOSOR470367-5GEA 1940 EUR
APOSOR470367-5G
4-(Aminomethyl)-4-(3,4-dichlorophenyl)cyclohexanol
4-(Aminomethyl)-4-(3,4-dichlorophenyl)cyclohexanol
Formel: C₁₃H₁₇Cl₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD06410570
CAS-Nummer: 887978-44-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR