1-Boc-3-(benzylamino)piperidine

Lieferant: Apollo Scientific

OR470368-5G
APOSOR470368-5GEA 1120 EUR
APOSOR470368-5G
1-Boc-3-(benzylamino)piperidine
1-Boc-3-(benzylamino)piperidine
Formel: C₁₇H₂₆N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06410626
CAS-Nummer: 183207-64-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR