6-Phenyl-2-piperidinecarboxylic acid

Lieferant: Apollo Scientific

OR470374-1G
APOSOR470374-1GEA 1230 EUR
APOSOR470374-1G
6-Phenyl-2-piperidinecarboxylic acid
6-Phenyl-2-piperidinecarboxylic acid
Formel: C₁₂H₁₅NO₂
Molecular Weight: 205,253 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410516
CAS-Nummer: 1219143-12-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR